Assay Design



Gene Transcripts

Gene Symbol
TMEM8B
Entrez Gene ID
51754
Gene Name
transmembrane protein 8B
Gene Aliases
C9orf127, NAG-5, NAG5, NGX6
Chromosome Location
Chr.9: 35825817 - 35865515 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001042589.2 NP_001036054.1 4 - 5 570 81
NM_001042590.2 NP_001036055.1 3 - 4 854 81
NM_016446.3 NP_057530.2 4 - 5 642 81
XM_005251483.4 XP_005251540.3 3 - 4 824 81
XM_011517901.2 XP_011516203.1 3 - 4 824 81
XM_011517902.2 XP_011516204.1 3 - 4 824 81
XM_011517903.2 XP_011516205.1 3 - 4 824 81
XM_011517904.2 XP_011516206.1 3 - 4 1025 81
XM_011517905.1 XP_011516207.1 3 - 4 824 81
XM_011517908.2 XP_011516210.1 2 - 3 277 81
XM_011517911.2 XP_011516213.1 4 - 5 570 81
XM_011517912.2 XP_011516214.1 3 - 4 416 81
XM_011517913.2 XP_011516215.1 3 - 4 631 81
XM_011517915.2 XP_011516217.1 2 - 3 453 81
GenBank mRNA AF070572.1 - 2 - 3 291 81
AF188239.1 - 4 - 5 603 81
BC082765.1 - 3 - 4 767 81
BC098265.1 - 1 - 2 185 81
BC098296.1 - 1 - 2 185 81
BC099733.1 - 1 - 2 185 81

View Full Product Details