Assay Design



Gene Transcripts

Gene Symbol
TMEM8B
Entrez Gene ID
51754
Gene Name
transmembrane protein 8B
Gene Aliases
C9orf127, NAG-5, NAG5, NGX6
Chromosome Location
Chr.9: 35825817 - 35865515 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001042589.2 NP_001036054.1 5 - 6 703 70
NM_001042590.2 NP_001036055.1 4 - 5 987 70
NM_016446.3 NP_057530.2 5 - 6 775 70
XM_005251483.4 XP_005251540.3 4 - 5 957 70
XM_011517901.2 XP_011516203.1 4 - 5 957 70
XM_011517902.2 XP_011516204.1 4 - 5 957 70
XM_011517903.2 XP_011516205.1 4 - 5 957 70
XM_011517904.2 XP_011516206.1 4 - 5 1158 70
XM_011517905.1 XP_011516207.1 4 - 5 957 70
XM_011517908.2 XP_011516210.1 3 - 4 410 70
XM_011517910.2 XP_011516212.1 3 - 4 288 70
XM_011517911.2 XP_011516213.1 5 - 6 703 70
XM_011517912.2 XP_011516214.1 4 - 5 549 70
XM_011517913.2 XP_011516215.1 4 - 5 764 70
XM_011517914.2 XP_011516216.1 2 - 3 209 70
XM_011517915.2 XP_011516217.1 3 - 4 586 70
XM_017014805.1 XP_016870294.1 2 - 3 348 70
XM_017014806.1 XP_016870295.1 3 - 4 400 70
GenBank mRNA AF070572.1 - 3 - 4 424 70
AF188239.1 - 5 - 6 736 70
AK074844.1 - 3 - 4 1098 70
BC082765.1 - 4 - 5 900 70
BC098265.1 - 2 - 3 318 70
BC098296.1 - 2 - 3 318 70
BC099733.1 - 2 - 3 318 70

View Full Product Details