Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001171195.1 | NP_001164666.1 | 5 - 6 | 968 | 97 |
NM_001171197.1 | NP_001164668.1 | 5 - 6 | 981 | 97 | |
XM_011517777.2 | XP_011516079.1 | 6 - 7 | 1374 | 97 | |
XM_017014416.1 | XP_016869905.1 | 6 - 7 | 877 | 97 | |
XM_017014424.1 | XP_016869913.1 | 5 - 6 | 1266 | 97 | |
XM_017014425.1 | XP_016869914.1 | 5 - 6 | 1074 | 97 | |
GenBank mRNA | BC030692.1 | - | 5 - 6 | 985 | 97 |
BC042393.1 | - | 5 - 6 | 971 | 97 |