Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018154.2 | NP_060624.1 | 1 - 2 | 281 | 67 |
GenBank mRNA | AB104486.1 | - | 1 - 2 | 109 | 67 |
AF279307.1 | - | 1 - 2 | 202 | 67 | |
AK001288.1 | - | 1 - 2 | 240 | 67 | |
AK001466.1 | - | 1 - 2 | 272 | 67 | |
AK223080.1 | - | 1 - 2 | 235 | 67 | |
AK302134.1 | - | 1 - 2 | 242 | 67 | |
BC007726.2 | - | 1 - 2 | 205 | 67 | |
BC010014.2 | - | 1 - 2 | 215 | 67 | |
BC036521.1 | - | 1 - 2 | 245 | 67 | |
CN426445.1 | - | 1 - 2 | 616 | 67 | |
CR457235.1 | - | 1 - 2 | 109 | 67 |