Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271043.2 | NP_001257972.1 | 5 - 6 | 900 | 84 |
NM_001271044.2 | NP_001257973.1 | 5 - 6 | 860 | 84 | |
NM_002501.3 | NP_002492.2 | 5 - 6 | 891 | 84 | |
XM_005259917.4 | XP_005259974.1 | 5 - 6 | 1178 | 84 | |
XM_005259918.4 | XP_005259975.1 | 5 - 6 | 898 | 84 | |
XM_005259919.3 | XP_005259976.1 | 5 - 6 | 1015 | 84 | |
XM_005259920.3 | XP_005259977.1 | 5 - 6 | 1577 | 84 | |
XM_006722760.2 | XP_006722823.1 | 5 - 6 | 1015 | 84 | |
XM_011528040.1 | XP_011526342.1 | 5 - 6 | 898 | 84 | |
XM_017026837.1 | XP_016882326.1 | 5 - 6 | 1149 | 84 | |
GenBank mRNA | AK295290.1 | - | 5 - 6 | 900 | 84 |
AK297261.1 | - | - | 859 | 84 | |
BC117113.1 | - | 4 - 5 | 854 | 84 | |
BC117115.1 | - | 4 - 5 | 854 | 84 | |
BT019732.1 | - | 5 - 6 | 823 | 84 | |
BX537794.1 | - | 5 - 6 | 897 | 84 | |
FJ861276.1 | - | 4 - 5 | 799 | 84 | |
L31881.1 | - | 4 - 5 | 859 | 84 | |
U18759.1 | - | 5 - 6 | 840 | 84 | |
U18760.1 | - | 4 - 5 | 481 | 84 | |
U18761.1 | - | 4 - 5 | 906 | 84 |