Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_057175.3 | NP_476516.1 | 16 - 17 | 2304 | 113 |
XM_005263236.2 | XP_005263293.1 | 16 - 17 | 2359 | 113 | |
GenBank mRNA | AF327722.1 | - | 16 - 17 | 2327 | 113 |
AJ314788.1 | - | 16 - 17 | 2256 | 113 | |
AK304417.1 | - | 15 - 16 | 1985 | 113 | |
AK314626.1 | - | 16 - 17 | 2359 | 113 | |
AY039242.1 | - | 16 - 17 | 2329 | 113 | |
AY112670.1 | - | 16 - 17 | 2048 | 113 | |
BC093928.1 | - | 16 - 17 | 2113 | 113 | |
BC104806.1 | - | 16 - 17 | 2113 | 113 |