Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_057175.3 | NP_476516.1 | 18 - 19 | 2563 | 88 |
XM_005263236.2 | XP_005263293.1 | 18 - 19 | 2615 | 88 | |
GenBank mRNA | AF327722.1 | - | 18 - 19 | 2586 | 88 |
AJ314788.1 | - | 18 - 19 | 2515 | 88 | |
AK304417.1 | - | 17 - 18 | 2244 | 88 | |
AK314626.1 | - | 18 - 19 | 2618 | 88 | |
AY039242.1 | - | 18 - 19 | 2588 | 88 | |
AY112670.1 | - | 18 - 19 | 2307 | 88 | |
BC093928.1 | - | 18 - 19 | 2372 | 88 | |
BC104806.1 | - | 18 - 19 | 2372 | 88 |