Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_057175.3 | NP_476516.1 | 2 - 3 | 390 | 144 |
XM_005263236.2 | XP_005263293.1 | 2 - 3 | 445 | 144 | |
GenBank mRNA | AF327722.1 | - | 2 - 3 | 413 | 144 |
AJ314788.1 | - | 2 - 3 | 342 | 144 | |
AK023387.1 | - | 2 - 3 | 111 | 144 | |
AK023402.1 | - | 2 - 3 | 413 | 144 | |
AK314626.1 | - | 2 - 3 | 445 | 144 | |
AY039242.1 | - | 2 - 3 | 415 | 144 | |
AY112670.1 | - | 2 - 3 | 134 | 144 | |
BC032642.1 | - | 2 - 3 | 415 | 144 | |
BC039818.1 | - | 2 - 3 | 405 | 144 | |
BC093928.1 | - | 2 - 3 | 199 | 144 | |
BC104806.1 | - | 2 - 3 | 199 | 144 |