Assay Design



Gene Transcripts

Gene Symbol
CNDP1
Entrez Gene ID
84735
Gene Name
carnosine dipeptidase 1
Gene Aliases
CN1, CPGL2, HsT2308
Chromosome Location
Chr.18: 74534457 - 74585025 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032649.5 NP_116038.4 6 - 7 963 91
GenBank mRNA AK098369.1 - 7 - 8 742 91
AK289916.1 - 6 - 7 818 91
AK303708.1 - 6 - 7 772 91
AK308716.1 - 6 - 7 916 91
AK314572.1 - 6 - 7 854 91
AY358756.1 - 6 - 7 952 91
BC110295.1 - 6 - 7 802 91
BC113512.1 - 6 - 7 791 91
BC117122.1 - 6 - 7 794 91

View Full Product Details