Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004432.3 | NP_004423.2 | 6 - 7 | 1033 | 103 |
XM_011517774.2 | XP_011516076.1 | 7 - 8 | 1434 | 103 | |
XM_017014408.1 | XP_016869897.1 | 8 - 9 | 1435 | 103 | |
XM_017014409.1 | XP_016869898.1 | 7 - 8 | 1073 | 103 | |
XM_017014410.1 | XP_016869899.1 | 7 - 8 | 1217 | 103 | |
XM_017014413.1 | XP_016869902.1 | 8 - 9 | 1381 | 103 | |
XM_017014414.1 | XP_016869903.1 | 7 - 8 | 1404 | 103 | |
XM_017014415.1 | XP_016869904.1 | 6 - 7 | 850 | 103 | |
XM_017014417.1 | XP_016869906.1 | 7 - 8 | 1294 | 103 | |
XM_017014419.1 | XP_016869908.1 | 6 - 7 | 1019 | 103 | |
XM_017014420.1 | XP_016869909.1 | 6 - 7 | 1022 | 103 | |
XM_017014421.1 | XP_016869910.1 | 6 - 7 | 1140 | 103 | |
XM_017014422.1 | XP_016869911.1 | 6 - 7 | 1309 | 103 | |
XM_017014423.1 | XP_016869912.1 | 6 - 7 | 966 | 103 | |
GenBank mRNA | AB209294.1 | - | 6 - 7 | 1033 | 103 |