Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001170750.1 | NP_001164221.1 | 8 - 9 | 1281 | 102 |
NM_001170751.1 | NP_001164222.1 | 8 - 9 | 1164 | 102 | |
NM_006307.4 | NP_006298.1 | 9 - 10 | 1341 | 102 | |
GenBank mRNA | AK125542.1 | - | 8 - 9 | 1262 | 102 |
AK289430.1 | - | 9 - 10 | 1288 | 102 | |
AK298802.1 | - | 8 - 9 | 1100 | 102 | |
BC020684.1 | - | 9 - 10 | 1296 | 102 | |
EU668359.1 | - | 9 - 10 | 1241 | 102 | |
U61374.1 | - | 9 - 10 | 1241 | 102 | |
U78093.1 | - | 9 - 10 | 1248 | 102 |