Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006946.2 | NP_008877.1 | 11 - 12 | 1425 | 90 |
XM_005274192.4 | XP_005274249.1 | 12 - 13 | 2087 | 90 | |
XM_005274193.3 | XP_005274250.1 | 11 - 12 | 1861 | 90 | |
XM_006718669.3 | XP_006718732.1 | 10 - 11 | 3774 | 90 | |
XM_006718671.3 | XP_006718734.1 | 12 - 13 | 1487 | 90 | |
XM_011545216.2 | XP_011543518.1 | 10 - 11 | 3784 | 90 | |
XM_011545217.2 | XP_011543519.1 | 10 - 11 | 3784 | 90 | |
XM_017018174.1 | XP_016873663.1 | 12 - 13 | 2179 | 90 | |
XM_017018175.1 | XP_016873664.1 | 12 - 13 | 1808 | 90 | |
XM_017018176.1 | XP_016873665.1 | 12 - 13 | 1528 | 90 | |
XM_017018177.1 | XP_016873666.1 | 11 - 12 | 1717 | 90 | |
XM_017018178.1 | XP_016873667.1 | 11 - 12 | 1400 | 90 | |
GenBank mRNA | AB008567.1 | - | 11 - 12 | 1425 | 90 |
BC139788.1 | - | 11 - 12 | 1396 | 90 |