Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006946.2 | NP_008877.1 | 13 - 14 | 1877 | 100 |
XM_005274192.4 | XP_005274249.1 | 14 - 15 | 2539 | 100 | |
XM_005274193.3 | XP_005274250.1 | 13 - 14 | 2313 | 100 | |
XM_006718669.3 | XP_006718732.1 | 12 - 13 | 4226 | 100 | |
XM_006718671.3 | XP_006718734.1 | 14 - 15 | 1939 | 100 | |
XM_011545216.2 | XP_011543518.1 | 12 - 13 | 4236 | 100 | |
XM_011545217.2 | XP_011543519.1 | 12 - 13 | 4236 | 100 | |
XM_017018174.1 | XP_016873663.1 | 14 - 15 | 2631 | 100 | |
XM_017018175.1 | XP_016873664.1 | 14 - 15 | 2260 | 100 | |
XM_017018176.1 | XP_016873665.1 | 14 - 15 | 1980 | 100 | |
XM_017018177.1 | XP_016873666.1 | 13 - 14 | 2169 | 100 | |
XM_017018178.1 | XP_016873667.1 | 13 - 14 | 1852 | 100 | |
GenBank mRNA | AB008567.1 | - | 13 - 14 | 1877 | 100 |