Assay Design
Gene Transcripts
- Gene Symbol
- MAPK10
- Entrez Gene ID
-
5602
- Gene Name
- mitogen-activated protein kinase 10
- Gene Aliases
-
JNK3, JNK3A, PRKM10, SAPK1b, p493F12, p54bSAPK
- Chromosome Location
-
Chr.4: 86012296 - 86594117 on Build GRCh38
View Full Product Details