Assay Design



Gene Transcripts

Gene Symbol
EBF1
Entrez Gene ID
1879
Gene Name
early B-cell factor 1
Gene Aliases
COE1, EBF, O/E-1, OLF1
Chromosome Location
Chr.5: 158695915 - 159099762 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001290360.2 NP_001277289.1 4 - 5 691 61
NM_001324101.1 NP_001311030.1 4 - 5 691 61
NM_001324103.1 NP_001311032.1 4 - 5 691 61
NM_001324106.1 NP_001311035.1 4 - 5 691 61
NM_001324107.1 NP_001311036.1 4 - 5 691 61
NM_001324108.1 NP_001311037.1 4 - 5 691 61
NM_001324109.1 NP_001311038.1 4 - 5 691 61
NM_001324111.1 NP_001311040.1 4 - 5 786 61
NM_024007.4 NP_076870.1 4 - 5 691 61
NM_182708.2 NP_874367.1 4 - 5 691 61
XM_017009192.1 XP_016864681.1 4 - 5 691 61
XM_017009193.1 XP_016864682.1 4 - 5 679 61
XM_017009194.1 XP_016864683.1 4 - 5 691 61
XM_017009195.1 XP_016864684.1 4 - 5 691 61
XM_017009196.1 XP_016864685.1 4 - 5 1626 61
XM_017009197.1 XP_016864686.1 4 - 5 1132 61
XM_017009198.1 XP_016864687.1 4 - 5 1051 61
XM_017009202.1 XP_016864691.1 4 - 5 691 61
XM_017009203.1 XP_016864692.1 4 - 5 691 61
GenBank mRNA AK289712.1 - 4 - 5 678 61
AK303698.1 - 2 - 3 317 61
AK304438.1 - 4 - 5 394 61
BC038805.1 - 4 - 5 702 61
BC041178.1 - 4 - 5 618 61

View Full Product Details