Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001139466.1 | NP_001132938.1 | 9 - 10 | 1254 | 69 |
NM_001139467.1 | NP_001132939.1 | 8 - 9 | 1109 | 69 | |
NM_001139468.1 | NP_001132940.1 | 9 - 10 | 1244 | 69 | |
NM_005647.3 | NP_005638.1 | 9 - 10 | 1373 | 69 | |
XM_011545571.2 | XP_011543873.1 | 8 - 9 | 1095 | 69 | |
GenBank mRNA | AB209608.1 | - | 4 - 5 | 1327 | 69 |
AK289409.1 | - | 8 - 9 | 1109 | 69 | |
AK290962.1 | - | 9 - 10 | 1237 | 69 | |
BC032708.1 | - | 9 - 10 | 1220 | 69 | |
BC052304.1 | - | 9 - 10 | 1373 | 69 | |
Y12781.1 | - | 9 - 10 | 1532 | 69 |