Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001207013.1 | NP_001193942.1 | 7 - 8 | 967 | 98 |
NM_001308287.1 | NP_001295216.1 | 5 - 6 | 793 | 98 | |
NM_002828.3 | NP_002819.2 | 6 - 7 | 898 | 98 | |
NM_080422.2 | NP_536347.1 | 6 - 7 | 898 | 98 | |
NM_080423.2 | NP_536348.1 | 6 - 7 | 898 | 98 | |
XM_005258124.3 | XP_005258181.1 | 7 - 8 | 965 | 98 | |
XM_005258125.3 | XP_005258182.1 | 7 - 8 | 965 | 98 | |
XM_011525705.2 | XP_011524007.1 | 8 - 9 | 1185 | 98 | |
XM_011525706.2 | XP_011524008.1 | 5 - 6 | 686 | 98 | |
XM_011525707.2 | XP_011524009.1 | 2 - 3 | 293 | 98 | |
XM_017025884.1 | XP_016881373.1 | 6 - 7 | 914 | 98 | |
XM_017025885.1 | XP_016881374.1 | 7 - 8 | 1100 | 98 | |
XM_017025886.1 | XP_016881375.1 | 5 - 6 | 788 | 98 | |
XM_017025887.1 | XP_016881376.1 | 5 - 6 | 739 | 98 | |
XM_017025888.1 | XP_016881377.1 | 5 - 6 | 739 | 98 | |
GenBank mRNA | AB209569.1 | - | 5 - 6 | 825 | 98 |
AK225706.1 | - | 7 - 8 | 846 | 98 | |
AK290649.1 | - | 6 - 7 | 780 | 98 | |
AK292570.1 | - | 6 - 7 | 798 | 98 | |
BC008244.1 | - | 6 - 7 | 741 | 98 | |
BC016727.1 | - | 6 - 7 | 810 | 98 | |
M25393.1 | - | 6 - 7 | 764 | 98 |