Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271043.2 | NP_001257972.1 | 8 - 9 | 1328 | 91 |
XM_005259917.4 | XP_005259974.1 | 7 - 8 | 1483 | 91 | |
XM_005259918.4 | XP_005259975.1 | 8 - 9 | 1326 | 91 | |
XM_005259920.3 | XP_005259977.1 | 8 - 9 | 2005 | 91 | |
XM_011528040.1 | XP_011526342.1 | 8 - 9 | 1326 | 91 | |
GenBank mRNA | AK295290.1 | - | 8 - 9 | 1328 | 91 |
FJ861276.1 | - | 7 - 8 | 1227 | 91 | |
U18760.1 | - | 7 - 8 | 909 | 91 |