Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001782.2 | NP_001773.1 | 2 - 3 | 314 | 101 |
XM_006716893.2 | XP_006716956.1 | 2 - 3 | 314 | 101 | |
XM_006716894.3 | XP_006716957.1 | 3 - 4 | 687 | 101 | |
XM_011518074.2 | XP_011516376.1 | 3 - 4 | 732 | 101 | |
XM_017015351.1 | XP_016870840.1 | 3 - 4 | 687 | 101 | |
GenBank mRNA | AF283777.2 | - | 2 - 3 | 252 | 101 |
AK307305.1 | - | 2 - 3 | 276 | 101 | |
AK309268.1 | - | 2 - 3 | 210 | 101 | |
AK310093.1 | - | 2 - 3 | 281 | 101 | |
AK310685.1 | - | 2 - 3 | 287 | 101 | |
AK311283.1 | - | 2 - 3 | 563 | 101 | |
BC011734.2 | - | 2 - 3 | 225 | 101 | |
BC030227.1 | - | 2 - 3 | 255 | 101 | |
DQ174771.1 | - | 3 - 4 | 683 | 101 | |
M54992.1 | - | 2 - 3 | 298 | 101 |