Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001782.2 | NP_001773.1 | 4 - 5 | 479 | 64 |
XM_006716893.2 | XP_006716956.1 | 4 - 5 | 479 | 64 | |
XM_006716894.3 | XP_006716957.1 | 5 - 6 | 852 | 64 | |
XM_011518074.2 | XP_011516376.1 | 5 - 6 | 897 | 64 | |
XM_017015351.1 | XP_016870840.1 | 5 - 6 | 852 | 64 | |
GenBank mRNA | AK310093.1 | - | 4 - 5 | 446 | 64 |
AK310685.1 | - | 4 - 5 | 452 | 64 | |
AK311283.1 | - | 4 - 5 | 728 | 64 | |
BC011734.2 | - | 4 - 5 | 390 | 64 | |
BC030227.1 | - | 4 - 5 | 420 | 64 | |
DQ174771.1 | - | 5 - 6 | 848 | 64 | |
M54992.1 | - | 4 - 5 | 463 | 64 |