Assay Design



Gene Transcripts

Gene Symbol
CHKA
Entrez Gene ID
1119
Gene Name
choline kinase alpha
Gene Aliases
CHK, CK, CKI, EK
Chromosome Location
Chr.11: 68052859 - 68121391 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_017017148.1 XP_016872637.1 1 - 2 2027 99

View Full Product Details