Assay Design



Gene Transcripts

Gene Symbol
SLC25A45
Entrez Gene ID
283130
Gene Name
solute carrier family 25 member 45
Gene Aliases
-
Chromosome Location
Chr.11: 65375192 - 65383701 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077241.2 NP_001070709.2 1 - 2 188 84
XM_017017571.1 XP_016873060.1 1 - 2 224 84
GenBank mRNA BC036869.1 - 1 - 2 151 84

View Full Product Details