Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016169.3 | NP_057253.2 | 11 - 12 | 1555 | 96 |
XM_011539858.2 | XP_011538160.1 | 12 - 13 | 1684 | 96 | |
XM_011539860.2 | XP_011538162.1 | 12 - 13 | 1681 | 96 | |
XM_011539861.2 | XP_011538163.1 | 11 - 12 | 1558 | 96 | |
XM_011539863.2 | XP_011538165.1 | 12 - 13 | 1419 | 96 | |
XM_017016323.1 | XP_016871812.1 | 12 - 13 | 1914 | 96 | |
GenBank mRNA | AF159447.1 | - | 11 - 12 | 1510 | 96 |
AF172319.1 | - | 11 - 12 | 1360 | 96 | |
AF175770.1 | - | 11 - 12 | 1530 | 96 | |
AK311503.1 | - | 5 - 6 | 594 | 96 | |
AK312319.1 | - | 11 - 12 | 1545 | 96 | |
BC013291.2 | - | 11 - 12 | 1531 | 96 |