Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001178133.1 | NP_001171604.1 | 4 - 5 | 787 | 67 |
NM_016169.3 | NP_057253.2 | 4 - 5 | 787 | 67 | |
XM_011539858.2 | XP_011538160.1 | 4 - 5 | 787 | 67 | |
XM_011539860.2 | XP_011538162.1 | 4 - 5 | 787 | 67 | |
XM_011539861.2 | XP_011538163.1 | 4 - 5 | 787 | 67 | |
XM_011539863.2 | XP_011538165.1 | 4 - 5 | 522 | 67 | |
XM_011539864.2 | XP_011538166.1 | 4 - 5 | 787 | 67 | |
XM_017016323.1 | XP_016871812.1 | 4 - 5 | 1017 | 67 | |
GenBank mRNA | AF144231.1 | - | 4 - 5 | 742 | 67 |
AF159447.1 | - | 4 - 5 | 742 | 67 | |
AF172319.1 | - | 4 - 5 | 592 | 67 | |
AF175770.1 | - | 4 - 5 | 762 | 67 | |
AF222345.1 | - | 4 - 5 | 670 | 67 | |
AK312319.1 | - | 4 - 5 | 777 | 67 | |
AY358550.1 | - | 4 - 5 | 742 | 67 | |
BC013291.2 | - | 4 - 5 | 763 | 67 |