Gene Symbol | Transcript Accession | Exon Location | Assay Location | Protein ID |
---|---|---|---|---|
FHOD1 | NM_001318202.1 | NP_001305131.1 | ||
NM_013241.2 | NP_037373.2 | |||
XM_006721180.1 | XP_006721243.1 | |||
XM_011523043.2 | XP_011521345.1 | |||
XM_011523044.1 | XP_011521346.1 | |||
XM_011523045.2 | XP_011521347.1 | |||
AA609867.1 | ||||
AB041046.1 | BAD06250.1 | |||
AB209584.1 | BAD92821.1 | |||
AF113615.1 | AAD39906.1 | |||
AK025992.1 | ||||
AK301157.1 | ||||
AY192154.1 | AAO38757.1 | |||
BC033084.1 | AAH33084.1 | |||
BU689676.1 | ||||
KJ658417.1 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv522852 | Chr16:67153308 - 68071571 on Build GRCh38 | Gain | KIAA0895L TMEM208 ENKD1 ATP6V0D1 AGRP FBXL8 E2F4 HSF4 LCAT NRN1L CTCF PARD6A NOL3 GFOD2 ELMO3 NUTF2 MIR328 KCTD19 PSKH1 CARMIL2 DPEP2 FHOD1 PLEKHG4 TRADD LRRC36 ZDHHC1 CTRL EDC4 HSD11B2 EXOC3L1 LOC105369155 ACD LRRC29 TSNAXIP1 SLC12A4 CENPT SLC9A5 LOC100131303 DPEP3 RANBP10 FAM65A LOC100505942 THAP11 DDX28 C16orf86 LOC107984813 DUS2 TPPP3 PSMB10 |
nsv827707 | Chr16:67148355 - 67289296 on Build GRCh38 | Loss | KIAA0895L TMEM208 NOL3 TRADD ELMO3 C16orf70 FBXL8 MIR328 E2F4 HSF4 EXOC3L1 LOC105369155 B3GNT9 LRRC29 FHOD1 PLEKHG4 SLC9A5 |
nsv952044 | Chr16:67141198 - 67244397 on Build GRCh38 | Deletion | KIAA0895L TMEM208 NOL3 TRADD ELMO3 C16orf70 FBXL8 MIR328 E2F4 HSF4 EXOC3L1 LOC105369155 B3GNT9 LRRC29 FHOD1 |
nsv457513 | Chr16:67181417 - 67282697 on Build GRCh38 | Loss | KIAA0895L EXOC3L1 TMEM208 ELMO3 LOC105369155 LRRC29 FHOD1 MIR328 PLEKHG4 E2F4 SLC9A5 |
nsv524492 | Chr16:67174030 - 67294550 on Build GRCh38 | Loss | KIAA0895L TMEM208 NOL3 ELMO3 MIR328 E2F4 KCTD19 EXOC3L1 LOC105369155 LRRC29 FHOD1 PLEKHG4 SLC9A5 |
nsv524363 | Chr16:67136529 - 68071571 on Build GRCh38 | Loss | KIAA0895L TMEM208 ENKD1 ATP6V0D1 AGRP FBXL8 E2F4 HSF4 LCAT NRN1L CTCF PARD6A NOL3 GFOD2 ELMO3 C16orf70 NUTF2 MIR328 KCTD19 PSKH1 CARMIL2 DPEP2 B3GNT9 FHOD1 PLEKHG4 TRADD LRRC36 ZDHHC1 CTRL EDC4 HSD11B2 EXOC3L1 LOC105369155 ACD LRRC29 TSNAXIP1 SLC12A4 CENPT SLC9A5 LOC100131303 DPEP3 RANBP10 FAM65A LOC100505942 THAP11 DDX28 C16orf86 LOC107984813 DUS2 TPPP3 PSMB10 |