Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000582.2 | NP_000573.1 | 1 - 2 | 151 | 63 |
NM_001040058.1 | NP_001035147.1 | 1 - 2 | 151 | 63 | |
NM_001040060.1 | NP_001035149.1 | 1 - 2 | 151 | 63 | |
NM_001251829.1 | NP_001238758.1 | 1 - 2 | 151 | 63 | |
NM_001251830.1 | NP_001238759.1 | 1 - 2 | 151 | 63 | |
GenBank mRNA | AF052124.1 | - | 1 - 2 | 73 | 63 |
AK057738.1 | - | 1 - 2 | 87 | 63 | |
AK075463.1 | - | 1 - 2 | 87 | 63 | |
AK290090.1 | - | 1 - 2 | 87 | 63 | |
AK290104.1 | - | 1 - 2 | 88 | 63 | |
AK295491.1 | - | 1 - 2 | 86 | 63 | |
AK296035.1 | - | 1 - 2 | 87 | 63 | |
AK315461.1 | - | 1 - 2 | 81 | 63 | |
AY956318.1 | - | 1 - 2 | 53 | 63 | |
BC007016.1 | - | 1 - 2 | 87 | 63 | |
BC017387.1 | - | 1 - 2 | 91 | 63 | |
BC022844.1 | - | 1 - 2 | 88 | 63 | |
BC093033.1 | - | 1 - 2 | 143 | 63 | |
BF699765.1 | - | 1 - 2 | 152 | 63 | |
BX648003.1 | - | 1 - 2 | 105 | 63 | |
CB117856.1 | - | 1 - 2 | 151 | 63 | |
DA419665.1 | - | 1 - 2 | 151 | 63 | |
J04765.1 | - | 1 - 2 | 94 | 63 | |
JF412667.1 | - | 1 - 2 | 149 | 63 | |
KR062183.1 | - | 1 - 2 | 86 | 63 | |
M83248.1 | - | 1 - 2 | 83 | 63 | |
X13694.1 | - | 1 - 2 | 53 | 63 |