Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_033274.4 | NP_150377.1 | 22 - 23 | 2830 | 110 |
XM_011534682.2 | XP_011532984.1 | 15 - 16 | 3586 | 110 | |
XM_017010009.1 | XP_016865498.1 | 15 - 16 | 2047 | 110 | |
GenBank mRNA | AF311317.1 | - | 22 - 23 | 2739 | 110 |
Y13786.2 | - | 22 - 23 | 2780 | 110 |