Assay Design



Gene Transcripts

Gene Symbol
ATP10A
Entrez Gene ID
57194
Gene Name
ATPase phospholipid transporting 10A (putative)
Gene Aliases
ATP10C, ATPVA, ATPVC
Chromosome Location
Chr.15: 25672241 - 25865144 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024490.3 NP_077816.1 11 - 12 2550 59
XM_005268261.4 XP_005268318.1 12 - 13 2680 59
XM_011521826.2 XP_011520128.1 12 - 13 2851 59
XM_011521828.2 XP_011520130.1 11 - 12 3279 59
XM_011521829.2 XP_011520131.1 8 - 9 1683 59
XM_011521830.2 XP_011520132.2 6 - 7 1353 59
XM_017022436.1 XP_016877925.1 11 - 12 2186 59
XM_017022437.1 XP_016877926.1 11 - 12 2550 59
GenBank mRNA AB011138.1 - 6 - 7 1438 59
AB051358.1 - 12 - 13 2643 59
BC052251.1 - 11 - 12 2556 59

View Full Product Details