Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001006109.1 | NP_001006109.1 | 3 - 4 | 595 | 72 |
NM_020187.2 | NP_064572.2 | 3 - 4 | 422 | 72 | |
XM_005247636.3 | XP_005247693.1 | 3 - 4 | 607 | 72 | |
XM_017006877.1 | XP_016862366.1 | 3 - 4 | 439 | 72 | |
GenBank mRNA | AF201934.1 | - | 3 - 4 | 435 | 72 |
AK025883.1 | - | 2 - 3 | 267 | 72 | |
AK309891.1 | - | 3 - 4 | 419 | 72 | |
BC009993.2 | - | 3 - 4 | 502 | 72 | |
BC010125.2 | - | 3 - 4 | 385 | 72 | |
BC050686.1 | - | 3 - 4 | 595 | 72 | |
BC088363.1 | - | 3 - 4 | 419 | 72 |