Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001166111.1 | NP_001159583.1 | 18 - 19 | 2341 | 105 |
NM_001166112.1 | NP_001159584.1 | 18 - 19 | 2279 | 105 | |
NM_001166113.1 | NP_001159585.1 | 19 - 20 | 2228 | 105 | |
NM_001166114.1 | NP_001159586.1 | 16 - 17 | 2160 | 105 | |
NM_006702.4 | NP_006693.3 | 19 - 20 | 2240 | 105 | |
GenBank mRNA | AJ004832.1 | - | 19 - 20 | 2114 | 105 |
AK294021.1 | - | 18 - 19 | 2298 | 105 | |
AK302462.1 | - | 18 - 19 | 2279 | 105 | |
BC038229.1 | - | 16 - 17 | 1977 | 105 | |
BC050553.1 | - | 19 - 20 | 2203 | 105 | |
BC051768.1 | - | 18 - 19 | 2146 | 105 |