Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001066.2 | NP_001057.1 | 4 - 5 | 547 | 110 |
XM_011542060.2 | XP_011540362.1 | 4 - 5 | 602 | 110 | |
XM_011542063.2 | XP_011540365.1 | 4 - 5 | 602 | 110 | |
XM_017002211.1 | XP_016857700.1 | 4 - 5 | 602 | 110 | |
XM_017002212.1 | XP_016857701.1 | 5 - 6 | 1386 | 110 | |
XM_017002213.1 | XP_016857702.1 | 6 - 7 | 1425 | 110 | |
XM_017002214.1 | XP_016857703.1 | 3 - 4 | 502 | 110 | |
XM_017002215.1 | XP_016857704.1 | 3 - 4 | 502 | 110 | |
GenBank mRNA | AY148473.1 | - | 4 - 5 | 464 | 110 |
BC011844.2 | - | 3 - 4 | 447 | 110 | |
BC042167.1 | - | 3 - 4 | 447 | 110 | |
BC052977.1 | - | 4 - 5 | 545 | 110 | |
BT019927.1 | - | 4 - 5 | 458 | 110 | |
M32315.1 | - | 4 - 5 | 547 | 110 | |
M55994.1 | - | 4 - 5 | 550 | 110 | |
S63368.1 | - | 3 - 4 | 350 | 110 |