Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001066.2 | NP_001057.1 | 9 - 10 | 1189 | 131 |
XM_017002215.1 | XP_016857704.1 | 8 - 9 | 1144 | 131 | |
GenBank mRNA | AY148473.1 | - | 7 - 8 | 993 | 131 |
BC011844.2 | - | 8 - 9 | 1089 | 131 | |
BC042167.1 | - | 8 - 9 | 1089 | 131 | |
BC052977.1 | - | 9 - 10 | 1187 | 131 | |
BT019927.1 | - | 9 - 10 | 1100 | 131 | |
M32315.1 | - | 9 - 10 | 1189 | 131 | |
M35857.1 | - | 6 - 7 | 755 | 131 | |
M55994.1 | - | 9 - 10 | 1192 | 131 | |
S63368.1 | - | 8 - 9 | 992 | 131 |