Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001112736.1 | NP_001106207.1 | 15 - 16 | 2717 | 82 |
NM_015224.3 | NP_056039.2 | 8 - 9 | 2068 | 82 | |
XM_005264999.1 | XP_005265056.1 | 15 - 16 | 2852 | 82 | |
XM_006713077.2 | XP_006713140.1 | 15 - 16 | 2852 | 82 | |
XM_006713078.2 | XP_006713141.1 | 9 - 10 | 1706 | 82 | |
XM_011533552.2 | XP_011531854.1 | 15 - 16 | 2852 | 82 | |
XM_011533553.2 | XP_011531855.1 | 14 - 15 | 2273 | 82 | |
XM_017006030.1 | XP_016861519.1 | 15 - 16 | 2627 | 82 | |
XM_017006031.1 | XP_016861520.1 | 15 - 16 | 2852 | 82 | |
XM_017006032.1 | XP_016861521.1 | 14 - 15 | 2273 | 82 | |
GenBank mRNA | AF180425.2 | - | 8 - 9 | 2352 | 82 |
AF273053.1 | - | 2 - 3 | 1052 | 82 | |
AK307599.1 | - | 7 - 8 | 1530 | 82 | |
BC070096.1 | - | 15 - 16 | 2715 | 82 | |
BC129987.1 | - | 7 - 8 | 1493 | 82 | |
BX648955.1 | - | 14 - 15 | 2199 | 82 |