Assay Design



Gene Transcripts

Gene Symbol
FAM208A
Entrez Gene ID
23272
Gene Name
family with sequence similarity 208 member A
Gene Aliases
C3orf63, RAP140, se89-1
Chromosome Location
Chr.3: 56620132 - 56683242 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001112736.1 NP_001106207.1 4 - 5 746 99
XM_005264999.1 XP_005265056.1 4 - 5 881 99
XM_006713077.2 XP_006713140.1 4 - 5 881 99
XM_011533552.2 XP_011531854.1 4 - 5 881 99
XM_011533553.2 XP_011531855.1 4 - 5 427 99
XM_017006030.1 XP_016861519.1 4 - 5 656 99
XM_017006031.1 XP_016861520.1 4 - 5 881 99
XM_017006032.1 XP_016861521.1 4 - 5 427 99
GenBank mRNA AK094486.1 - 4 - 5 876 99
BC070096.1 - 4 - 5 744 99
BX648955.1 - 3 - 4 228 99

View Full Product Details