Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_153267.4 | NP_694999.3 | 4 - 5 | 1119 | 89 |
NR_125850.1 | 4 - 5 | 1119 | 89 | ||
XM_011518471.1 | XP_011516773.1 | 4 - 5 | 1181 | 89 | |
XM_011518475.2 | XP_011516777.1 | 4 - 5 | 1181 | 89 | |
XM_017014587.1 | XP_016870076.1 | 4 - 5 | 1181 | 89 | |
GenBank mRNA | AK304040.1 | - | 4 - 5 | 882 | 89 |
AK304531.1 | - | 4 - 5 | 707 | 89 | |
BC016383.1 | - | 4 - 5 | 837 | 89 | |
BC063634.1 | - | 4 - 5 | 1096 | 89 |