Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_153267.4 | NP_694999.3 | 8 - 9 | 1755 | 90 |
NR_125850.1 | 8 - 9 | 1755 | 90 | ||
XM_011518471.1 | XP_011516773.1 | 8 - 9 | 1817 | 90 | |
XM_011518475.2 | XP_011516777.1 | 8 - 9 | 1817 | 90 | |
XM_017014587.1 | XP_016870076.1 | 8 - 9 | 1817 | 90 | |
GenBank mRNA | AK304040.1 | - | 8 - 9 | 1515 | 90 |
AK304531.1 | - | 8 - 9 | 1343 | 90 | |
BC016383.1 | - | 8 - 9 | 1473 | 90 | |
BC063634.1 | - | 8 - 9 | 1732 | 90 |