Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_028393.1 | 7 - 8 | 1841 | 140 | |
NR_028394.1 | 8 - 9 | 1969 | 140 | ||
NR_028395.1 | 8 - 9 | 1969 | 140 | ||
NR_028396.1 | 7 - 8 | 1841 | 140 | ||
NR_028397.1 | 7 - 8 | 1706 | 140 | ||
NR_028398.1 | 5 - 6 | 1460 | 140 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001012957.1 | NP_001012975.1 | 3 - 4 | 1173 | 140 |
NM_001012959.1 | NP_001012977.1 | 3 - 4 | 1173 | 140 | |
NM_001164538.1 | NP_001158010.1 | 3 - 4 | 1173 | 140 | |
NM_001164539.1 | NP_001158011.1 | 3 - 4 | 1173 | 140 | |
NM_001164540.1 | NP_001158012.1 | 3 - 4 | 1173 | 140 | |
NM_001164541.1 | NP_001158013.1 | 3 - 4 | 1173 | 140 | |
NM_001164542.1 | NP_001158014.1 | 3 - 4 | 1173 | 140 | |
NM_001164544.1 | NP_001158016.1 | 3 - 4 | 1173 | 140 | |
NM_001164545.1 | NP_001158017.1 | 3 - 4 | 1173 | 140 | |
NM_001164546.1 | NP_001158018.1 | 3 - 4 | 1173 | 140 | |
NM_001164547.1 | NP_001158019.1 | 3 - 4 | 1173 | 140 | |
NM_001164548.1 | NP_001158020.1 | 3 - 4 | 1173 | 140 | |
NM_001164549.1 | NP_001158021.1 | 3 - 4 | 1173 | 140 | |
NM_001164551.1 | NP_001158023.1 | 3 - 4 | 1173 | 140 | |
NM_018662.2 | NP_061132.2 | 3 - 4 | 1173 | 140 | |
GenBank mRNA | AB007926.1 | - | 3 - 4 | 1159 | 140 |
AF222980.1 | - | 3 - 4 | 1173 | 140 | |
AJ506177.1 | - | 3 - 4 | 1173 | 140 | |
BC151225.1 | - | 3 - 4 | 1159 | 140 | |
FJ804176.1 | - | 3 - 4 | 1173 | 140 | |
FJ804177.1 | - | 3 - 4 | 1173 | 140 | |
FJ804178.1 | - | 3 - 4 | 1173 | 140 | |
FJ804181.1 | - | 3 - 4 | 1173 | 140 | |
FJ804183.1 | - | 3 - 4 | 1173 | 140 | |
FJ804184.1 | - | 3 - 4 | 1173 | 140 | |
FJ804185.1 | - | 3 - 4 | 1173 | 140 | |
FJ804186.1 | - | 3 - 4 | 1173 | 140 | |
FJ804187.1 | - | 3 - 4 | 1173 | 140 | |
FJ804188.1 | - | 3 - 4 | 1173 | 140 | |
FJ804189.1 | - | 3 - 4 | 1173 | 140 | |
FJ804190.1 | - | 3 - 4 | 1173 | 140 | |
FJ804193.1 | - | 3 - 4 | 1173 | 140 | |
FJ804194.1 | - | 3 - 4 | 1173 | 140 | |
FJ804195.1 | - | 4 - 5 | 1241 | 140 | |
FJ804196.1 | - | 3 - 4 | 1173 | 140 | |
FJ804199.1 | - | 3 - 4 | 1173 | 140 | |
FJ804201.1 | - | 3 - 4 | 1173 | 140 | |
FJ804202.1 | - | 3 - 4 | 1173 | 140 | |
FJ804205.1 | - | 3 - 4 | 1173 | 140 | |
FJ804206.1 | - | 3 - 4 | 1173 | 140 | |
FJ804207.1 | - | 3 - 4 | 1173 | 140 | |
FJ804211.1 | - | 3 - 4 | 1173 | 140 |