Assay Design



Gene Transcripts

Gene Symbol
CD3G
Entrez Gene ID
917
Gene Name
CD3g molecule
Gene Aliases
CD3-GAMMA, IMD17, T3G
Chromosome Location
Chr.11: 118344307 - 118353782 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000073.2 NP_000064.1 2 - 3 158 105
XM_005271724.3 XP_005271781.1 2 - 3 195 105
XM_006718941.2 XP_006719004.1 2 - 3 146 105
GenBank mRNA AK223314.1 - 2 - 3 111 105
AK313966.1 - 2 - 3 146 105
BC113830.1 - 2 - 3 104 105
X04145.1 - 2 - 3 115 105

View Full Product Details