Assay Design



Gene Transcripts

Gene Symbol
CD3G
Entrez Gene ID
917
Gene Name
CD3g molecule
Gene Aliases
CD3-GAMMA, IMD17, T3G
Chromosome Location
Chr.11: 118344307 - 118353782 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000073.2 NP_000064.1 3 - 4 391 77
XM_005271724.3 XP_005271781.1 3 - 4 428 77
XM_006718941.2 XP_006719004.1 3 - 4 379 77
GenBank mRNA AK223314.1 - 3 - 4 344 77
AK310578.1 - 4 - 5 519 77
AK310622.1 - 4 - 5 549 77
AK313966.1 - 3 - 4 379 77
AK315895.1 - 3 - 4 335 77
BC113830.1 - 3 - 4 337 77
X04145.1 - 3 - 4 348 77

View Full Product Details