Assay Design



Gene Transcripts

Gene Symbol
CTRC
Entrez Gene ID
11330
Gene Name
chymotrypsin C
Gene Aliases
CLCR, ELA4
Chromosome Location
Chr.1: 15438442 - 15449247 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007272.2 NP_009203.2 7 - 8 811 61
XM_011540550.1 XP_011538852.1 6 - 7 665 61
GenBank mRNA AK289447.1 - 7 - 8 810 61
BC015118.1 - 7 - 8 817 61
BC059779.1 - 6 - 7 644 61
CR749296.1 - 4 - 5 475 61
S82198.1 - 7 - 8 788 61
Y13697.1 - 6 - 7 740 61

View Full Product Details