Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000186.3 | NP_000177.2 | 14 - 15 | 2477 | 195 |
GenBank mRNA | AK226113.1 | - | 9 - 10 | 4263 | 195 |
AK291395.1 | - | 14 - 15 | 2316 | 195 | |
BC142699.1 | - | 14 - 15 | 2289 | 195 | |
M17517.1 | - | 3 - 4 | 516 | 195 | |
Y00716.1 | - | 14 - 15 | 2310 | 195 |