Assay Design



Gene Transcripts

Gene Symbol
MCOLN3
Entrez Gene ID
55283
Gene Name
mucolipin 3
Gene Aliases
TRP-ML3, TRPML3
Chromosome Location
Chr.1: 85018082 - 85048902 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018298.10 NP_060768.8 3 - 4 550 112
XM_005271003.1 XP_005271060.1 4 - 5 947 112
XM_006710750.1 XP_006710813.1 4 - 5 766 112
XM_011541739.2 XP_011540041.2 3 - 4 474 112
XM_011541741.2 XP_011540043.1 3 - 4 481 112
GenBank mRNA AF475085.1 - 3 - 4 456 112
AK292206.1 - 3 - 4 482 112
AK314354.1 - 3 - 4 510 112
BC060765.1 - 4 - 5 780 112

View Full Product Details