Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001253693.1 | NP_001240622.1 | 7 - 8 | 922 | 121 |
NM_018298.10 | NP_060768.8 | 8 - 9 | 1090 | 121 | |
XM_005271003.1 | XP_005271060.1 | 9 - 10 | 1487 | 121 | |
XM_006710750.1 | XP_006710813.1 | 9 - 10 | 1306 | 121 | |
XM_011541739.2 | XP_011540041.2 | 8 - 9 | 1014 | 121 | |
XM_011541740.2 | XP_011540042.1 | 7 - 8 | 848 | 121 | |
XM_011541741.2 | XP_011540043.1 | 8 - 9 | 1021 | 121 | |
XM_011541742.1 | XP_011540044.1 | 9 - 10 | 1224 | 121 | |
GenBank mRNA | AF475085.1 | - | 8 - 9 | 996 | 121 |
AK001868.1 | - | 7 - 8 | 828 | 121 | |
AK095148.1 | - | 2 - 3 | 965 | 121 | |
AK292206.1 | - | 8 - 9 | 1022 | 121 | |
AK314354.1 | - | 8 - 9 | 1050 | 121 | |
BX647497.1 | - | 10 - 11 | 1243 | 121 |