Assay Design



Gene Transcripts

Gene Symbol
ODF2
Entrez Gene ID
4957
Gene Name
outer dense fiber of sperm tails 2
Gene Aliases
CT134, ODF2/1, ODF2/2, ODF84
Chromosome Location
Chr.9: 128455155 - 128501292 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001242352.1 NP_001229281.1 12 - 13 1660 61
NM_001242353.1 NP_001229282.1 12 - 13 1746 61
NM_001242354.1 NP_001229283.1 11 - 12 1432 61
NM_002540.4 NP_002531.3 14 - 15 1948 61
NM_153432.1 NP_702910.1 11 - 12 1506 61
NM_153433.1 NP_702911.1 12 - 13 1673 61
NM_153435.1 NP_702913.1 12 - 13 1566 61
NM_153436.1 NP_702914.1 11 - 12 1330 61
NM_153437.2 NP_702915.1 12 - 13 1596 61
NM_153439.1 NP_702917.1 11 - 12 1449 61
NM_153440.1 NP_702918.1 11 - 12 1376 61
XM_005252010.2 XP_005252067.1 11 - 12 1458 61
XM_005252011.2 XP_005252068.1 11 - 12 1398 61
XM_005252012.3 XP_005252069.1 12 - 13 1648 61
XM_005252013.3 XP_005252070.1 12 - 13 1595 61
XM_005252014.3 XP_005252071.1 12 - 13 1644 61
XM_005252017.4 XP_005252074.1 12 - 13 1549 61
XM_005252018.2 XP_005252075.1 13 - 14 1754 61
XM_005252019.3 XP_005252076.1 13 - 14 1773 61
XM_005252022.2 XP_005252079.1 12 - 13 1572 61
XM_005252023.2 XP_005252080.1 12 - 13 1543 61
XM_005252024.3 XP_005252081.1 12 - 13 1562 61
XM_005252026.4 XP_005252083.1 12 - 13 1491 61
XM_005252029.2 XP_005252086.1 12 - 13 1572 61
XM_006717126.3 XP_006717189.1 12 - 13 1648 61
XM_011518722.2 XP_011517024.1 12 - 13 1572 61
XM_011518723.2 XP_011517025.1 12 - 13 1515 61
XM_011518724.2 XP_011517026.1 13 - 14 1736 61
XM_011518725.2 XP_011517027.1 13 - 14 1740 61
XM_011518726.2 XP_011517028.1 13 - 14 1758 61
XM_011518728.2 XP_011517030.1 12 - 13 1509 61
XM_011518729.2 XP_011517031.1 12 - 13 1445 61
XM_011518730.2 XP_011517032.1 12 - 13 1572 61
XM_011518731.2 XP_011517033.1 11 - 12 1401 61
XM_011518732.2 XP_011517034.1 12 - 13 1358 61
XM_011518733.2 XP_011517035.1 11 - 12 1344 61
XM_011518744.1 XP_011517046.1 11 - 12 1398 61
XM_017014764.1 XP_016870253.1 13 - 14 1729 61
XM_017014765.1 XP_016870254.1 13 - 14 1748 61
XM_017014766.1 XP_016870255.1 11 - 12 1458 61
XM_017014767.1 XP_016870256.1 13 - 14 1740 61
XM_017014768.1 XP_016870257.1 12 - 13 1532 61
XM_017014769.1 XP_016870258.1 12 - 13 1561 61
XM_017014770.1 XP_016870259.1 13 - 14 1716 61
XM_017014771.1 XP_016870260.1 13 - 14 1697 61
XM_017014772.1 XP_016870261.1 13 - 14 1773 61
XM_017014773.1 XP_016870262.1 13 - 14 1749 61
XM_017014774.1 XP_016870263.1 12 - 13 1459 61
XM_017014775.1 XP_016870264.1 13 - 14 1716 61
XM_017014776.1 XP_016870265.1 13 - 14 1697 61
XM_017014777.1 XP_016870266.1 12 - 13 1499 61
XM_017014778.1 XP_016870267.1 12 - 13 1602 61
XM_017014779.1 XP_016870268.1 11 - 12 1343 61
XM_017014780.1 XP_016870269.1 12 - 13 1526 61
XM_017014781.1 XP_016870270.1 11 - 12 1281 61
GenBank mRNA AK093367.1 - 11 - 12 1456 61
AK299303.1 - 12 - 13 1508 61
AK300793.1 - 11 - 12 1281 61
AK301754.1 - 11 - 12 1373 61
AK301842.1 - 11 - 12 1298 61
AK301855.1 - 11 - 12 1352 61
AK302684.1 - 12 - 13 1600 61
AK308278.1 - 10 - 11 1212 61
AY319414.1 - 11 - 12 1482 61
AY366499.1 - 12 - 13 1443 61
BC010629.1 - 14 - 15 1746 61
BC091500.1 - 12 - 13 1560 61
DQ444713.1 - 10 - 11 1273 61
DQ444714.1 - 10 - 11 1216 61

View Full Product Details