Assay Design



Gene Transcripts

Gene Symbol
ODF2
Entrez Gene ID
4957
Gene Name
outer dense fiber of sperm tails 2
Gene Aliases
CT134, ODF2/1, ODF2/2, ODF84
Chromosome Location
Chr.9: 128455155 - 128501292 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001242352.1 NP_001229281.1 17 - 18 2287 70
NM_001242353.1 NP_001229282.1 17 - 18 2373 70
NM_002540.4 NP_002531.3 19 - 20 2575 70
NM_153433.1 NP_702911.1 17 - 18 2300 70
NM_153435.1 NP_702913.1 17 - 18 2193 70
XM_005252010.2 XP_005252067.1 16 - 17 2085 70
XM_005252011.2 XP_005252068.1 16 - 17 2025 70
XM_005252012.3 XP_005252069.1 17 - 18 2275 70
XM_005252013.3 XP_005252070.1 17 - 18 2222 70
XM_005252014.3 XP_005252071.1 17 - 18 2271 70
XM_005252017.4 XP_005252074.1 17 - 18 2176 70
XM_005252018.2 XP_005252075.1 18 - 19 2381 70
XM_005252019.3 XP_005252076.1 18 - 19 2400 70
XM_005252022.2 XP_005252079.1 17 - 18 2199 70
XM_005252023.2 XP_005252080.1 17 - 18 2170 70
XM_005252024.3 XP_005252081.1 17 - 18 2189 70
XM_005252026.4 XP_005252083.1 17 - 18 2118 70
XM_005252029.2 XP_005252086.1 17 - 18 2199 70
XM_006717126.3 XP_006717189.1 17 - 18 2275 70
XM_011518722.2 XP_011517024.1 17 - 18 2199 70
XM_011518723.2 XP_011517025.1 17 - 18 2142 70
XM_011518724.2 XP_011517026.1 18 - 19 2363 70
XM_011518725.2 XP_011517027.1 18 - 19 2367 70
XM_011518726.2 XP_011517028.1 18 - 19 2385 70
XM_011518728.2 XP_011517030.1 17 - 18 2136 70
XM_011518729.2 XP_011517031.1 17 - 18 2072 70
XM_011518730.2 XP_011517032.1 17 - 18 2199 70
XM_011518731.2 XP_011517033.1 16 - 17 2028 70
XM_011518732.2 XP_011517034.1 17 - 18 1985 70
XM_011518733.2 XP_011517035.1 16 - 17 1971 70
XM_017014764.1 XP_016870253.1 18 - 19 2356 70
XM_017014765.1 XP_016870254.1 18 - 19 2375 70
XM_017014766.1 XP_016870255.1 16 - 17 2085 70
XM_017014767.1 XP_016870256.1 18 - 19 2367 70
XM_017014768.1 XP_016870257.1 17 - 18 2159 70
XM_017014769.1 XP_016870258.1 17 - 18 2188 70
XM_017014770.1 XP_016870259.1 18 - 19 2343 70
XM_017014771.1 XP_016870260.1 18 - 19 2324 70
XM_017014772.1 XP_016870261.1 18 - 19 2400 70
XM_017014773.1 XP_016870262.1 18 - 19 2376 70
XM_017014774.1 XP_016870263.1 17 - 18 2086 70
XM_017014775.1 XP_016870264.1 18 - 19 2343 70
XM_017014776.1 XP_016870265.1 18 - 19 2324 70
XM_017014777.1 XP_016870266.1 17 - 18 2126 70
XM_017014778.1 XP_016870267.1 17 - 18 2229 70
XM_017014779.1 XP_016870268.1 16 - 17 1970 70
XM_017014780.1 XP_016870269.1 17 - 18 2153 70
XM_017014781.1 XP_016870270.1 16 - 17 1908 70
GenBank mRNA AK299303.1 - 17 - 18 2135 70
AK302684.1 - 17 - 18 2227 70
AY366499.1 - 17 - 18 2070 70
BC010629.1 - 19 - 20 2373 70
BC091500.1 - 17 - 18 2187 70
DQ444713.1 - 15 - 16 1900 70
DQ444714.1 - 15 - 16 1843 70

View Full Product Details