Assay Design



Gene Transcripts

Gene Symbol
ODF2
Entrez Gene ID
4957
Gene Name
outer dense fiber of sperm tails 2
Gene Aliases
CT134, ODF2/1, ODF2/2, ODF84
Chromosome Location
Chr.9: 128455155 - 128501292 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001242352.1 NP_001229281.1 19 - 20 2546 59
NM_001242353.1 NP_001229282.1 19 - 20 2632 59
NM_002540.4 NP_002531.3 21 - 22 2834 59
NM_153433.1 NP_702911.1 19 - 20 2559 59
NM_153435.1 NP_702913.1 19 - 20 2452 59
XM_005252010.2 XP_005252067.1 18 - 19 2500 59
XM_005252011.2 XP_005252068.1 18 - 19 2440 59
XM_005252012.3 XP_005252069.1 19 - 20 2690 59
XM_005252013.3 XP_005252070.1 19 - 20 2637 59
XM_005252014.3 XP_005252071.1 19 - 20 2686 59
XM_005252017.4 XP_005252074.1 19 - 20 2591 59
XM_005252018.2 XP_005252075.1 20 - 21 2796 59
XM_005252019.3 XP_005252076.1 20 - 21 2815 59
XM_005252022.2 XP_005252079.1 19 - 20 2614 59
XM_005252023.2 XP_005252080.1 19 - 20 2585 59
XM_005252024.3 XP_005252081.1 19 - 20 2604 59
XM_005252026.4 XP_005252083.1 19 - 20 2533 59
XM_005252029.2 XP_005252086.1 19 - 20 2458 59
XM_006717126.3 XP_006717189.1 19 - 20 2534 59
XM_011518722.2 XP_011517024.1 19 - 20 2614 59
XM_011518723.2 XP_011517025.1 19 - 20 2557 59
XM_011518724.2 XP_011517026.1 20 - 21 2778 59
XM_011518725.2 XP_011517027.1 20 - 21 2782 59
XM_011518726.2 XP_011517028.1 20 - 21 2800 59
XM_011518728.2 XP_011517030.1 19 - 20 2551 59
XM_011518729.2 XP_011517031.1 19 - 20 2487 59
XM_011518730.2 XP_011517032.1 19 - 20 2458 59
XM_011518731.2 XP_011517033.1 18 - 19 2443 59
XM_011518732.2 XP_011517034.1 19 - 20 2400 59
XM_011518733.2 XP_011517035.1 18 - 19 2386 59
XM_017014764.1 XP_016870253.1 20 - 21 2771 59
XM_017014765.1 XP_016870254.1 20 - 21 2790 59
XM_017014766.1 XP_016870255.1 18 - 19 2344 59
XM_017014767.1 XP_016870256.1 20 - 21 2626 59
XM_017014768.1 XP_016870257.1 19 - 20 2574 59
XM_017014769.1 XP_016870258.1 19 - 20 2603 59
XM_017014770.1 XP_016870259.1 20 - 21 2758 59
XM_017014771.1 XP_016870260.1 20 - 21 2739 59
XM_017014772.1 XP_016870261.1 20 - 21 2659 59
XM_017014773.1 XP_016870262.1 20 - 21 2635 59
XM_017014774.1 XP_016870263.1 19 - 20 2345 59
XM_017014775.1 XP_016870264.1 20 - 21 2602 59
XM_017014776.1 XP_016870265.1 20 - 21 2583 59
XM_017014777.1 XP_016870266.1 19 - 20 2385 59
XM_017014778.1 XP_016870267.1 19 - 20 2488 59
XM_017014779.1 XP_016870268.1 18 - 19 2229 59
XM_017014780.1 XP_016870269.1 19 - 20 2412 59
XM_017014781.1 XP_016870270.1 18 - 19 2167 59
GenBank mRNA AK126816.1 - 2 - 3 604 59
AK299303.1 - 19 - 20 2394 59
AK302684.1 - 19 - 20 2486 59
AY366499.1 - 19 - 20 2329 59
BC010629.1 - 21 - 22 2632 59
BC012785.2 - 2 - 3 209 59
BC091500.1 - 19 - 20 2446 59
DQ444713.1 - 17 - 18 2159 59
DQ444714.1 - 17 - 18 2102 59

View Full Product Details