Assay Design



Gene Transcripts

Gene Symbol
ODF2
Entrez Gene ID
4957
Gene Name
outer dense fiber of sperm tails 2
Gene Aliases
CT134, ODF2/1, ODF2/2, ODF84
Chromosome Location
Chr.9: 128455155 - 128501292 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001242352.1 NP_001229281.1 1 - 2 249 81
NM_001242354.1 NP_001229283.1 1 - 2 249 81
NM_153432.1 NP_702910.1 1 - 2 249 81
NM_153435.1 NP_702913.1 1 - 2 249 81
NM_153439.1 NP_702917.1 1 - 2 249 81
XM_005252010.2 XP_005252067.1 1 - 2 201 81
XM_005252011.2 XP_005252068.1 1 - 2 198 81
XM_005252017.4 XP_005252074.1 1 - 2 138 81
XM_005252026.4 XP_005252083.1 1 - 2 137 81
XM_011518722.2 XP_011517024.1 1 - 2 198 81
XM_011518723.2 XP_011517025.1 1 - 2 198 81
XM_011518730.2 XP_011517032.1 1 - 2 198 81
XM_011518731.2 XP_011517033.1 1 - 2 198 81
XM_011518733.2 XP_011517035.1 1 - 2 198 81
XM_011518744.1 XP_011517046.1 1 - 2 198 81
XM_017014766.1 XP_016870255.1 1 - 2 201 81
XM_017014777.1 XP_016870266.1 1 - 2 145 81
XM_017014779.1 XP_016870268.1 1 - 2 103 81
XM_017014781.1 XP_016870270.1 1 - 2 98 81
GenBank mRNA AK093367.1 - 1 - 2 200 81
AK295662.1 - 1 - 2 97 81
AK299303.1 - 1 - 2 97 81
AK300793.1 - 1 - 2 98 81
AK301754.1 - 1 - 2 173 81
AK309917.1 - 1 - 2 97 81
AY366499.1 - 1 - 2 126 81
BM563800.1 - - 173 81
DB094573.1 - 1 - 2 249 81

View Full Product Details