Assay Design



Gene Transcripts

Gene Symbol
ODF2
Entrez Gene ID
4957
Gene Name
outer dense fiber of sperm tails 2
Gene Aliases
CT134, ODF2/1, ODF2/2, ODF84
Chromosome Location
Chr.9: 128455155 - 128501292 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001242352.1 NP_001229281.1 2 - 3 343 82
NM_001242354.1 NP_001229283.1 2 - 3 343 82
XM_005252017.4 XP_005252074.1 2 - 3 232 82
XM_005252018.2 XP_005252075.1 3 - 4 437 82
XM_005252019.3 XP_005252076.1 3 - 4 456 82
XM_005252026.4 XP_005252083.1 2 - 3 231 82
XM_017014770.1 XP_016870259.1 3 - 4 456 82
XM_017014771.1 XP_016870260.1 3 - 4 437 82
XM_017014772.1 XP_016870261.1 3 - 4 456 82
XM_017014773.1 XP_016870262.1 3 - 4 432 82
XM_017014774.1 XP_016870263.1 2 - 3 142 82
XM_017014775.1 XP_016870264.1 3 - 4 456 82
XM_017014776.1 XP_016870265.1 3 - 4 437 82
XM_017014777.1 XP_016870266.1 2 - 3 239 82
XM_017014778.1 XP_016870267.1 3 - 4 456 82
XM_017014779.1 XP_016870268.1 2 - 3 197 82
XM_017014780.1 XP_016870269.1 3 - 4 437 82
XM_017014781.1 XP_016870270.1 2 - 3 192 82
GenBank mRNA AK295662.1 - 2 - 3 191 82
AK299303.1 - 2 - 3 191 82
AK300793.1 - 2 - 3 192 82
AK309917.1 - 2 - 3 191 82
BM928430.1 - 3 - 4 488 82

View Full Product Details