Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164317.1 | NP_001157789.1 | 23 - 24 | 4228 | 64 |
NM_001164318.1 | NP_001157790.1 | 23 - 24 | 4228 | 64 | |
NM_001164319.1 | NP_001157791.1 | 23 - 24 | 4228 | 64 | |
NM_001457.3 | NP_001448.2 | 23 - 24 | 4228 | 64 | |
XM_005264978.1 | XP_005265035.1 | 23 - 24 | 4208 | 64 | |
GenBank mRNA | AB191258.1 | - | 23 - 24 | 4206 | 64 |
AB209889.1 | - | 4 - 5 | 1097 | 64 | |
AB371580.1 | - | 23 - 24 | 4208 | 64 | |
AB371581.1 | - | 23 - 24 | 4206 | 64 | |
AB371582.1 | - | 23 - 24 | 4206 | 64 | |
AF042166.1 | - | 23 - 24 | 4194 | 64 | |
AF043045.1 | - | 23 - 24 | 4228 | 64 | |
BX641085.1 | - | 22 - 23 | 3786 | 64 | |
CR749793.1 | - | 22 - 23 | 3713 | 64 |