Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164317.1 | NP_001157789.1 | 32 - 33 | 5541 | 75 |
NM_001164318.1 | NP_001157790.1 | 31 - 32 | 5415 | 75 | |
NM_001164319.1 | NP_001157791.1 | 30 - 31 | 5376 | 75 | |
NM_001457.3 | NP_001448.2 | 31 - 32 | 5448 | 75 | |
XM_005264978.1 | XP_005265035.1 | 31 - 32 | 5449 | 75 | |
GenBank mRNA | AB191258.1 | - | 31 - 32 | 5393 | 75 |
AB209889.1 | - | 12 - 13 | 2317 | 75 | |
AB371580.1 | - | 32 - 33 | 5521 | 75 | |
AB371581.1 | - | 30 - 31 | 5354 | 75 | |
AB371582.1 | - | 31 - 32 | 5426 | 75 | |
AF042166.1 | - | 31 - 32 | 5414 | 75 | |
AF043045.1 | - | 31 - 32 | 5448 | 75 | |
AL833551.1 | - | 5 - 6 | 1551 | 75 | |
BX641085.1 | - | 30 - 31 | 4973 | 75 | |
CR749793.1 | - | 29 - 30 | 4861 | 75 |